Learn More
APOE Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a recombinant APOE.
Supplier: Abnova Corporation PAB8547
Description
Chylomicron remnants and very low density lipoprotein (VLDL) remnants are rapidly removed from the circulation by receptor-mediated endocytosis in the liver. Apolipoprotein E, a main apoprotein of the chylomicron, binds to a specific receptor on liver cells and peripheral cells. ApoE is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. The APOE gene is mapped to chromosome 19 in a cluster with APOC1 and APOC2. Defects in apolipoprotein E result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. [provided by RefSeq
Specifications
APOE | |
Polyclonal | |
Rabbit polyclonal antibody raised against a recombinant APOE. | |
In PBS, pH 7.2 (30% glycerol, 0.01% thimerosal) | |
AD2/LPG/MGC1571/apoprotein | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
APOE | |
APOE | |
Recombinant protein corresponding to human APOE. | |
RUO | |
348 | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only