Learn More
APOH, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant APOH
Supplier: Abnova Corporation PAB28473
Description
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections. [provided by RefSeq
Sequence: ECREVKCPFPSRPDNGFVNYPAKPTLYYKDKATFGCHDGYSLDGPEEIECTKLGNWSAMPSCKASCKVPVKKATVVYQGERVKIQEKFKNGMLHGDKVSFFCKNKEKKCSYTEDAQCIDGTIEVPKCFKEHSSLAFSKTDASDVKSpecifications
APOH | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant APOH | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
P02749 | |
APOH | |
Recombinant protein corresponding to amino acids of human APOH | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry(1:50-1:200) Western Blot(1:100-1:250) The optimal working dilution should be determined by the end user. | |
APOH | |
B2G1/BG | |
Rabbit | |
Antigen affinity purification | |
RUO | |
350 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only