Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP fibrils, Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89130450 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-450 Supplier Abnova Corporation Supplier No. PAB28923
Add to Cart
Add to Cart

Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.

This gene encodes a cell surface receptor and transmembrane precursor protein that is cleaved by secretases to form a number of peptides. Some of these peptides are secreted and can bind to the acetyltransferase complex APBB1/TIP60 to promote transcriptional activation, while others form the protein basis of the amyloid plaques found in the brains of patients with Alzheimer disease. Mutations in this gene have been implicated in autosomal dominant Alzheimer disease and cerebroarterial amyloidosis (cerebral amyloid angiopathy). Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen APP fibrils
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.
Dilution Dot blot (1:1000) ELISA Immunocytochemistry Immunohistochemistry Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation PBS pH 7.4, 50% glycerol and 0.09% sodium azide
Gene APP
Gene Accession No. P05067
Gene Alias AAA/ABETA/ABPP/AD1/APPI/CTFgamma/CVAP/PN2
Gene Symbols APP
Host Species Rabbit
Immunogen Fibrils prepared from human Abeta42 peptide.
Purification Method Protein A affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351
Test Specificity PAB28923 recognizes generic epitopes common to many amyloid fibrils and fibrillar oligomers, but not prefibrilllar oligomers or natively folded proteins. 1:1000 dilution of PAB28923 was sufficient for detection of amyloid oligomers in 10ug of mouse brain lysates by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.