Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aqp1, Rabbit, APC, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Rat Aqp1.
Supplier: Abnova Corporation PAB28848
Description
Sequence: MASEFKKKLFSpecifications
Aqp1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant Rat Aqp1. | |
PBS pH 7.4, 50% glycerol and 0.09% sodium azide | |
NP_036910.1 | |
Aqp1 | |
Recombinant protein corresponding to Aqp1 N-terminus. | |
100 μg | |
Primary | |
0.5 ug/mL of PAB28848 was sufficient for detection of Aqp1, ∼28.5kDa and somtimes larger glycosylated bands ∼35-50kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
APC | |
Immunofluorescence (1:200-400) Western Blot (1:1000-4000) The optimal working dilution should be determined by the end user. | |
Aqp1 | |
CHIP28 | |
Rabbit | |
Affinity purification | |
RUO | |
25240 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction