Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqp2, Rabbit, PerCP, Polyclonal Antibody, Abnova™

Catalog No. 89130396 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-396 Supplier Abnova Corporation Supplier No. PAB28871
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Rat Aqp2.

Sequence: CLKGLEPDTDWEEREVRRRQ

Specifications

Antigen Aqp2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate PerCP
Description Rabbit polyclonal antibody raised against recombinant Rat Aqp2.
Dilution Immunofluorescence (1:400) Western Blot (1:1000-4000) The optimal working dilution should be determined by the end user.
Formulation PBS pH 7.4, 50% glycerol and 0.09% sodium azide
Gene Aqp2
Gene Accession No. NP_037041.2
Gene Alias AQP-2/MGC156502/aquaporin-2
Gene Symbols Aqp2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Aqp2 C-terminus.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25386
Test Specificity 0.5 ug/mL of PAB28871 was sufficient for detection of Aqp2, ∼28.8kDa and sometimes larger glycosylated bands ∼35-50kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.