Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqp2 (phospho S269), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118876 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-876 Supplier Abnova Corporation Supplier No. PAB17203
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Aqp2.

Specifications

Antigen Aqp2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of Aqp2.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 0.1mg/mL BSA)
Gene Aqp2
Gene Accession No. P34080
Gene Alias AQP-2/MGC156502/aquaporin-2
Gene Symbols Aqp2
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S269 of rat Aqp2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25386
Test Specificity This antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that This antibody will react with bovine, canine and mouse tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen. specific to the ∼29k AQP2 protein phosphorylated at Ser269. Also recognizes the glycosylated form of AQP2 at ∼ 37k. Immunolabeling of the AQP2 band is blocked by preadsorption with the phospho-peptide used as antigen but not by the corresponding dephospho-peptide.
Target Species Bovine, Canine, Mouse, Rat
Content And Storage For long term store at -80°C is recommended, but shorter term storage at -20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50 % glycerol.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.