Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqp2 Rabbit anti-Bovine, Human, Mouse, Pig, Rat, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89115611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-611 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-611 Supplier Abnova Corporation Supplier No. PAB12114
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Aqp2.

Specifications

Antigen Aqp2
Applications Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Aqp2.
Dilution Immunocytochemistry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.01% BSA, 1mM sodium azide)
Gene Aqp2
Gene Accession No. P34080
Gene Alias AQP-2/MGC156502/aquaporin-2
Gene Symbols Aqp2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-300 of rat Aqp2.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25386
Target Species Bovine, Human, Mouse, Pig, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.