Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQP9 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-398 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-398 Supplier Abnova Corporation Supplier No. PAB18839
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of AQP9.

The aquaporins are a family of water-selective membrane channels. The protein encoded by this gene allows passage of a wide variety of noncharged solutes. It stimulates urea transport and osmotic water permeability; there are contradicting reports about its role in providing glycerol permeability. The encoded protein may also play a role in specialized leukocyte functions such as immunological response and bactericidal activity. [provided by RefSeq

Specifications

Antigen AQP9
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of AQP9.
Dilution ELISA (1:40000-1:80000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide, 2% BSA
Gene AQP9
Gene Accession No. NM_020980
Gene Alias HsT17287/SSC1
Gene Symbols AQP9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at N-terminus of human AQP9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.