Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP12, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115041 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-041 Supplier Abnova Corporation Supplier No. PAB1136
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of human ARHGAP12.

RHO GTPases (see ARHA, MIM 165390) that participate in a wide variety of cellular processes are active in their GTP-bound form and inactive in their GDP-bound form. The hydrolysis of GTP and the inactivation of these GTPases are regulated by GTPase-activating proteins (GAPs), such as ARHGAP12 (Zhang et al., 2002 [PubMed 11854031]).[supplied by OMIM

Sequence: ILVKKTNDDWWQVKPD

Specifications

Antigen ARHGAP12
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of human ARHGAP12.
Dilution ELISA (1:100-1:1000) Western Blot (2.0 ug/mL) The optimal dilution should be determined by the end user.
Formulation In serum (50% glycerol, 0.025% sodium azide)
Gene ARHGAP12
Gene Accession No. NP_060757
Gene Alias DKFZp779N2050/FLJ10971/FLJ20737/FLJ21785/FLJ45709
Gene Symbols ARHGAP12
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 40-56 of human ARHGAP12.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94134
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.