Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP21, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124738 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-738 Supplier Abnova Corporation Supplier No. PAB23039
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ARHGAP21.

ARHGAP21 functions preferentially as a GTPase-activating protein (GAP) for CDC42 (MIM 116952) and regulates the ARP2/3 complex (MIM 604221) and F-actin dynamics at the Golgi through control of CDC42 activity (Dubois et al., 2005 [PubMed 15793564]).[supplied by OMIM

Sequence: VPPCLTTSAPLIRRQLSHDHESVGPPSLDAQPNSKTERSKSYDEGLDDYREDAKLSFKHVSSLKGIKIADSQKSSEDSGSRKDSSSEVFSDAAKEG

Specifications

Antigen ARHGAP21
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ARHGAP21.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ARHGAP21
Gene Accession No. Q5T5U3
Gene Alias ARHGAP10/DKFZp761L0424/FLJ33323/FLJ90108
Gene Symbols ARHGAP21
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ARHGAP21.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57584
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.