Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF15 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89126046 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-046 Supplier Abnova Corporation Supplier No. PAB24336

Rabbit polyclonal antibody raised against recombinant ARHGEF15.

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein-coupled receptors. This gene encodes a protein that functions as a specific guanine nucleotide exchange factor for RhoA. It also interacts with ephrin A4 in vascular smooth muscle cells. [provided by RefSeq

Sequence: PTHRLLQASSLSDMQRWLGAFPTPGPLPCSPDTIYEDCDCSQELCSESSAPAKTEGRSLESRAAPKHLHKTPEGWLKGLPGAFPAQLVCEVTGEH

Specifications

Antigen ARHGEF15
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ARHGEF15.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ARHGEF15
Gene Accession No. O94989
Gene Alias ARGEF15/FLJ13791/KIAA0915/MGC44868/Vsm-RhoGEF
Gene Symbols ARHGEF15
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ARHGEF15.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22899
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.