Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arhgef17, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Arhgef17.
Supplier: Abnova Corporation PAB15715
Description
Sequence: MLAGSDAIIRQHKAACLRITALLVCAELLWVGTSAGVVLTIPTSPSTVSCPRAPLSPAGLCQGHTGHVRFLAAVQLPEGFNLLCSTPPPPPDTGPEKLPSLDHRDSPRRRGPTSARPKMLVISGGDGSEDFRLSSGGGGSSETVGRDDSTNHLLLWRVSpecifications
Arhgef17 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Arhgef17. | |
In PBS (50% glycerol, 0.02% sodium azide) | |
AK122250 | |
Arhgef17 | |
Recombinant GST fusion protein corresponding to 158 mouse Arhgef17. | |
RUO | |
207212 | |
Mouse | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Arhgef17 | |
8030463K16/AI428794/AW558066/BC035332/mKIAA0337 | |
Rabbit | |
50 μg | |
Primary | |
Specific to recombinant protein GX0072. This antibody detects mARHGEF17 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction