Learn More
ARL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ARL1.
Supplier: Abnova Corporation PAB4256
Description
The protein encoded by this gene belongs to the ARL (ADP-ribosylation factor-like) family of proteins, which are structurally related to ADP-ribosylation factors (ARFs). ARFs, described as activators of cholera toxin (CT) ADP-ribosyltransferase activity, regulate intracellular vesicular membrane trafficking, and stimulate a phospholipase D (PLD) isoform. Although, ARL proteins were initially thought not to activate CT or PLD, later work showed that they are weak stimulators of PLD and CT in a phospholipid dependent manner. [provided by RefSeq
Specifications
ARL1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ARL1. | |
In PBS (0.09% sodium azide) | |
NP_001168;P40616 | |
ARL1 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ARL1. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
ARL1 | |
ARFL1 | |
Rabbit | |
Protein G purification | |
RUO | |
400 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only