Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-ARL13B, Polyclonal,

Catalog No. ABN445MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN445MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN445MI Supplier MilliporeSigma™ Supplier No. ABN445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects ARL13B in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunohistochemistry

ARL13B/ARL2L1 is an important neuronal developmental protein, as well as, a cilium-specific protein required for the control of the microtubule-based ciliary axoneme structure. ARL13B/ARL2L1 is required for proper patterning in neural development during neural tube formation, and ARL13B/ARL2L1 is critical for cerebral cortex development and the initial polarization of radial glia scaffold, the first step in cortex development. ARL13B/ARL2L1 also controls the migration and placement of post mitotic interneurons. ARL13B/ARL2L1 is found in cellular projections and cilium and is widely expressed in the developing system as well in the gut. ARL13B/ARL2L1 also is a member of the small GTPase superfamily as it binds GTP but does not cleave it.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL13B
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q3SXY8
Gene Symbols ARL13B; ARL2L1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human ARL13B.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_878899
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.