Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL6IP1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116187 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-187 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-187 Supplier Abnova Corporation Supplier No. PAB12918
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ARL6IP1.

Specifications

Antigen ARL6IP1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ARL6IP1.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ARL6IP1
Gene Accession No. Q15041
Gene Alias AIP1/ARL6IP/ARMER/KIAA0069
Gene Symbols ARL6IP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 15 amino acids of human ARL6IP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23204
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.