Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNTL, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115860 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-860 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-860 Supplier Abnova Corporation Supplier No. PAB12484
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ARNTL.

The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen ARNTL
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ARNTL.
Dilution Western Blot (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ARNTL
Gene Accession No. O00327
Gene Alias BMAL1/BMAL1c/JAP3/MGC47515/MOP3/PASD3/TIC/bHLHe5
Gene Symbols ARNTL
Host Species Rabbit
Immunogen Recombinant protein corresponding to human ARNTL.
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 406
Test Specificity This antibody is specific to MOP3/BMAL1.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.