Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Arpin, Polyclonal,

Catalog No. ABT251MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT251MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT251MI Supplier MilliporeSigma™ Supplier No. ABT251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Arpin in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Arpin (UniProt Q7Z6K5; also known as Arp2/3 inhibition protein, UPF0552 protein C15orf38) is encoded by the ARPIN (also known as C15orf38) gene (Gene ID 348110) in human. Cell migration requires the generation of branched actin networks that power the protrusion of the plasma membrane in lamellipodia. The actin-related proteins 2 and 3 (Arp2/3) complex nucleates actin filaments by binding to the side of an existing filament and initiating branch formation. Arpin functions as a negative regulator of Arp2/3 activity, allowing cells to control directional persistence of migration by fine-tuning actin nucleation activity at the leading edge of the lamellipodium. Arpin depletion in both mammalian cells and Dictyostelium discoideum amoeba is reported to result in straighter trajectories. Arpin microinjection is demonstrated to induce fish keratocytes to turn.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arpin
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q7Z6K5
Gene Symbols ARPIN; C15orf38
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Arpin near the C-terminus.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_872422
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.