Learn More
Description
Sulfatases (EC 3.1.5.6), such as ARSI, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 [PubMed 16174644]).[supplied by OMIM
Specifications
Specifications
| Antigen | ARSI |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of ARSI. |
| Dilution | Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:60000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) |
| Gene | ARSI |
| Gene Accession No. | Q5FYB1 |
| Gene Alias | FLJ16069 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.