Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arx, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127026 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-026 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-026 Supplier Abnova Corporation Supplier No. PAB2539

Rabbit polyclonal antibody raised against synthetic peptide of Arx.

Specifications

Antigen Arx
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description aristaless related homeobox
Formulation In PBS (0.09% sodium azide)
Gene Arx
Gene Accession No. NP_031518;O35085
Gene Symbols Arx
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of mouse Arx.
Purification Method Protein A purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11878
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.