Learn More
ASAH2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ASAH2.
Supplier: Abnova Corporation PAB13057
Description
Ceramidases (EC 3.5.1.23), such as ASAH2, catalyze hydrolysis of the N-acyl linkage of ceramide, a second messenger in a variety of cellular events, to produce sphingosine. Sphingosine exerts both mitogenic and apoptosis-inducing activities, and its phosphorylated form functions as an intra- and intercellular second messenger (see MIM 603730) (Mitsutake et al., 2001 [PubMed 11328816]).[supplied by OMIM
Specifications
ASAH2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ASAH2. | |
In PBS (0.02% sodium azide) | |
NP_063946 | |
ASAH2 | |
A synthetic peptide corresponding to C-terminus 17 amino acids of human ASAH2. | |
RUO | |
56624 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
ASAH2 | |
HNAC1/MGC129777/NCDase | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only