Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asap1 Rabbit anti-Bovine, Chicken, Chimpanzee, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114618 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-618 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-618 Supplier Abnova Corporation Supplier No. PAB10088
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Asap1.

Specifications

Antigen Asap1
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Asap1.
Dilution ELISA (1:2000-1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Asap1
Gene Accession No. NP_034156(seealsoAAC98349);Q9QWY8
Gene Alias AV239055/Ddef1/PAP/mKIAA1249/s19
Gene Symbols Asap1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 777-787 of mouse Asap1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13196
Test Specificity Reactivity occurs against both the phosphorylated and non-phosphorylated forms of the protein at residue Y782.
Target Species Bovine, Canine, Chicken, Chimpanzee, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.