Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASGR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89122079 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-079 Supplier Abnova Corporation Supplier No. PAB20573
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ASGR1.

Partially deglycosylated plasma glycoproteins and immunoglobulin IgA2 allotypes are efficiently and specifically removed from circulation by a receptor-mediated process. The asialoglycoprotein receptor binds to desialylated (galactosyl-terminal) glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociate. Then the receptor is recycled back to the cell surface and the ligand is transported to the lysosomes for degradation. [provided by RefSeq

Sequence: MTKEYQDLQHLDNEESDHHQLRKGPPPPQPLLQRLCSGPRL

Specifications

Antigen ASGR1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ASGR1.
Dilution Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ASGR1
Gene Accession No. P07306
Gene Alias HL-1/ASGPR/ASGPR1/CLEC4H1
Gene Symbols ASGR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ASGR1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 432
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.