Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATAD5 Rabbit anti-Chimpanzee, Dog, Human, Polyclonal Antibody, Abnova™

Catalog No. 89114704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-704 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-704 Supplier Abnova Corporation Supplier No. PAB10222
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ATAD5.

Specifications

Antigen ATAD5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ATAD5.
Dilution ELISA (1:3000-1:15000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene ATAD5
Gene Accession No. Q96QE3;CAC44537
Gene Alias C17orf41/FLJ12735/FRAG1
Gene Symbols ATAD5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 63-76 of human ATAD5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79915
Test Specificity The following proteins should be considered to be highly homologous to Elg1 when using this antibody: human chromosome fragility associated gene 1 (NP_079133), hypothetical dog protein FLJ12735 (XP_548276), hypothetical chimpanzee protein FLJ12735 (XP_511388), and a hypothetical human protein of 1224 residues (CAH10412). All show 100% homology to the immunogen.
Target Species Canine, Chimpanzee, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.