Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG10 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114379 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-379 Supplier Abnova Corporation Supplier No. PAB0732
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ATG10.

Autophagy is a process for the bulk degradation of cytosolic compartments by lysosomes. ATG10 is an E2-like enzyme involved in 2 ubiquitin-like modifications essential for autophagosome formation: ATG12 (MIM 609608)-ATG5 (MIM 604261) conjugation and modification of a soluble form of MAP-LC3 (MAP1LC3A; MIM 601242), a homolog of yeast Apg8, to a membrane-bound form (Nemoto et al., 2003 [PubMed 12890687]).[supplied by OMIM

Specifications

Antigen ATG10
Applications Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ATG10.
Dilution Immunohistochemistry (1:100-1:1000) Immunofluorescence (1:100-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene ATG10
Gene Accession No. Q9H0Y0
Gene Alias APG10/APG10L/DKFZp586I0418/FLJ13954/pp12616
Gene Symbols ATG10
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human ATG10.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83734
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.