Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG12 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114363 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-363 Supplier Abnova Corporation Supplier No. PAB0713
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ATG12.

Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. ATG12 is the human homolog of a yeast protein involved in autophagy (Mizushima et al., 1998 [PubMed 9852036]).[supplied by OMIM

Specifications

Antigen ATG12
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ATG12.
Dilution Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene ATG12
Gene Accession No. O94817
Gene Alias APG12/APG12L/FBR93/HAPG12
Gene Symbols ATG12
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human ATG12.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9140
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.