Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atg16l1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115647 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-647 Supplier Abnova Corporation Supplier No. PAB12149
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Atg16l1.

Specifications

Antigen Atg16l1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Atg16l1.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene Atg16l1
Gene Accession No. Q8C0J2
Gene Alias 1500009K01Rik/AI035639/AI225866/Apg16l/Atg16l/WDR30
Gene Symbols Atg16l1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of mouse Atg16l1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 77040
Target Species Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.