Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-375 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-375 Supplier Abnova Corporation Supplier No. PAB0727
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ATG5.

Specifications

Antigen ATG5
Applications Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ATG5.
Dilution Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene ATG5
Gene Accession No. Q9H1Y0
Gene Alias APG5/APG5-LIKE/APG5L/ASP/hAPG5
Gene Symbols ATG5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ATG5.
Purification Method Protein G purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9474
Target Species Human
Content And Storage Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.