Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115858 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-858 Supplier Abnova Corporation Supplier No. PAB12482
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ATG5.

Specifications

Antigen ATG5
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ATG5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene ATG5
Gene Accession No. Q9H1Y0
Gene Alias APG5/APG5-LIKE/APG5L/ASP/hAPG5
Gene Symbols ATG5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-50 of human ATG5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9474
Test Specificity This antibody is selective for the full-length and calpain cleaved isoform proteins. The short isoform is missing a.a. 1-79. The calpain cleaved form of ATG5 is missing a.a. 195-275.
Target Species Bovine, Fish, Human, Mouse, Pig, Primate, Rat, Xenopus species
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.