Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG9B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120786 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-786 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-786 Supplier Abnova Corporation Supplier No. PAB19255
Add to Cart
Add to Cart

Rabbit polyclonal antibody rasied against synthetic peptide of ATG9B.

Specifications

Antigen ATG9B
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody rasied against synthetic peptide of ATG9B.
Dilution Western Blot (1-2 ug/mL) Immunocytochemistry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ATG9B
Gene Accession No. NP_775952
Gene Alias APG9L2/NOS3AS/SONE
Gene Symbols ATG9B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids near C-terminus of human ATG9B.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285973
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.