Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP2C2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATP2C2.
Supplier: Abnova Corporation PAB14291
Specifications
ATP2C2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ATP2C2. | |
In PBS (50% glycerol, 0.01% sodium azide) | |
DKFZp686H22230/KIAA0703/SPCA2 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
ATP2C2 | |
ATP2C2 | |
A synthetic peptide corresponding to amino acids 426-438 of human ATP2C2. | |
RUO | |
9914 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction