Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AURKA Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120401 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-401 Supplier Abnova Corporation Supplier No. PAB18841
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of AURKA.

The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen AURKA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of AURKA.
Dilution ELISA (1:10000-1:40000) Western Blot (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide, 2% BSA
Gene AURKA
Gene Accession No. O14965
Gene Alias AIK/ARK1/AURA/AURORA2/BTAK/MGC34538/STK15/STK6/STK7
Gene Symbols AURKA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids of human AURKA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6790
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.