Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora Kinase A/B/C, Rabbit, Clone: SP201, Abnova™

Catalog No. 89110310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-310 Supplier Abnova Corporation Supplier No. MAB12169

Rabbit monoclonal antibody raised against synthetic peptide of human Aurora Kinase A/B/C.

The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen Aurora Kinase A/B/C
Applications Flow Cytometry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP201
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human Aurora Kinase A/B/C.
Dilution Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene AURKA
Gene Alias AIK/ARK1/AURA/AURORA2/BTAK/MGC34538/STK15/STK6/STK7
Gene Symbols AURKA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus region of human Aurora kinase A, B, and C.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6790
Test Specificity 48 kDa (Aurora A), 40 kDa (Aurora B), and 35 kDa (Aurora C).
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.