Learn More
BACE2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BACE2.
Supplier: Abnova Corporation PAB4706
Description
Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer's disease and a frequent complication of Down syndrome. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein by 2 proteases, one of which is the protein encoded by this gene. This gene localizes to the 'Down critical region' of chromosome 21. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease. Three transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Specifications
BACE2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BACE2. | |
In PBS (0.09% sodium azide) | |
Q9Y5Z0 | |
BACE2 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human BACE2. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user. | |
BACE2 | |
AEPLC/ALP56/ASP1/ASP21/BAE2/CDA13/CEAP1/DRAP | |
Rabbit | |
Protein G purification | |
RUO | |
25825 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only