Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BANP, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120845 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-845 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-845 Supplier Abnova Corporation Supplier No. PAB19312
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody rasied against synthetic peptide of BANP.

This gene encodes a protein that binds to matrix attachment regions. The protein functions as a tumor suppressor and cell cycle regulator. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Specifications

Antigen BANP
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody rasied against synthetic peptide of BANP.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene BANP
Gene Accession No. NP_001167014
Gene Alias BEND1/DKFZp761H172/FLJ10177/FLJ20538/SMAR1/SMARBP1
Gene Symbols BANP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids near internal region of human BANP.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54971
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.