Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-Bassoon, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABN255
Description
Specifically detects Bassoon in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Protein bassoon (also known as bassoon presynaptic cytomatrix protein, neuronal double zinc finger protein, zinc finger protein 231) is a cytoplasmic protein encoded by the BSN gene in human (also known as KIAA0434 and ZNF23118). Bassoon is found primarily in neurons in brain (PMID 9806829) and most likely functions as a scaffolding protein involved in organizing the presynaptic cytoskeleton. Murine and rodent orthologs are reported to be detected in axon terminals of hippocampal neurons, where it is highly concentrated in the vicinity of the active zone, indicating a potential involvement in cytomatrix organization at the site of neurotransmitter release (PMID 9679147).
Specifications
Bassoon | |
Polyclonal | |
O88737 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat |
Immunohistochemistry (Paraffin), Western Blot | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Bsn | |
GST-tagged recombinant protein corresponding to mouse Bassoon. | |
100 μL | |
Neuroscience | |
NP_031593 | |
Stable for one year at 2°C to 8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction