Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBC3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89115922 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-922 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-922 Supplier Abnova Corporation Supplier No. PAB12555
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BBC3.

Sequence: EQHLESPVPSAPGALAG

Specifications

Antigen BBC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BBC3.
Dilution Western Blot (1:500-1:1) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene BBC3
Gene Alias JFY1/PUMA
Gene Symbols BBC3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to exon-3-encoded region of human BBC3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27113
Test Specificity Reacts with a 17 residue sequence [EQHLESPVPSAPGALAG] found in the exon-3-encoded region in the human PUMA-alpha and PUMA-beta forms of the protein. Recognizes both forms of the protein.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.