Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcar1 (phospho Y762), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Bcar1.
Supplier: Abnova Corporation PAB7934
Specifications
Bcar1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Bcar1. | |
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
Cas/Crkas/P130CAS | |
Rabbit | |
100 μL | |
Primary | |
This rat sequence is the same as Tyr-664 in human p130 Cas and Tyr-668 in mouse p130 Cas. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
ELISA (1:2000) Immunoprecipitation (5-10 ul) Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
Bcar1 | |
Bcar1 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y762 of rat Bcar1. | |
RUO | |
25414 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction