Learn More
BCL2 (phospho T56), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BCL2.
Supplier: Abnova Corporation PAB12610
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Sequence: GHTPHSpecifications
BCL2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BCL2. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
Bcl-2 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes BCL2 with the phosphorylation site Thr56. It does not cross-react with non-phosphospecific peptide. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
BCL2 | |
BCL2 | |
Synthetic phosphopeptide corresponding to residues surrounding T56 of human BCL2. | |
RUO | |
596 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only