Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-690 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-690 Supplier Abnova Corporation Supplier No. PAB16877
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BCL2.

This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq

Sequence: AGRTGYDNREIVMKY

Specifications

Antigen BCL2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BCL2.
Dilution ELISA (1:100000-1:400000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene BCL2
Gene Alias Bcl-2
Gene Symbols BCL2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to BCL2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.