Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-BCL2A1/BFL-1, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABC490
Description
Specifically detects BCL2A1/BFL-1 in Human samples, and it is validated for Immunohistochemistry, Western Blotting
Bcl-2-related protein A1 (BCL2A1), also known as protein BFL-1, protein GRS, and Bcl-2-like protein 5 (Bcl2-L-5), is a member of the Bcl-2 family. BCL2A1/BFL-1 can interact with Bax and suppress apoptosis by inhibiting the release of cytochrome c and caspase-3 activation. BCL2A1/BFL-1 expression is reported in peripheral blood, spleen, bone marrow, lung, small intestine, and testis.
Specifications
| BCL2A1/BFL-1 | |
| Polyclonal | |
| Q16548 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human |
| Immunohistochemistry, Western Blot | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| BCL2A1; BCL2L5; BFL1; GRS; HBPA1 | |
| KLH-conjugated linear peptide corresponding to the near N-terminal of human BCL2A1/BFL-1. | |
| 100 μg | |
| Apoptosis, Cancer | |
| NP_001108207 | |
| Stable for one year at 2°C to 8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction