Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-BCL2L2, Polyclonal, Abnova™

Catalog No. 89115470 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-470 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-470 Supplier Abnova Corporation Supplier No. PAB11958

Specifications

Antigen BCL2L2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In BBS, pH 8.2
Gene Symbols BCL2L2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 16-29 of human BCL2L2.
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 599
Target Species Bovine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.