Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-BCL6, Polyclonal,

Catalog No. ABC1480MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC1480MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1480MI Supplier MilliporeSigma™ Supplier No. ABC1480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects BCL6 in Rat samples, and it is validated for Western Blotting

BCL6 Antibody: BCL6 was first identified as a proto-oncogene in diffuse large B cell lymphoma. BCL6, a transcriptional repressor, plays an important role in lymphomagenesis. It binds Stat recognition-like DNA elements and is required for affinity maturation of mature B cells in germinal centers and Th1/Th2 differentiation, a process that critically depends on BCL6-mediated transcriptional repression of p53. BCL6 is a pharmacological target for eradication of leukemia-initiating cells in chronic myeloid leukemia (CML).
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL6
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.
Gene Accession No. P41182
Gene Symbols BCL6; BCL5; LAZ3; ZBTB27; ZNF51
Host Species Rabbit
Immunogen BCL6 antibody was raised against an 18 amino acid synthetic peptide near the center of human BCL6.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Target Species Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.