Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCLAF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-852 Supplier Abnova Corporation Supplier No. PAB20352
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant BCLAF1.

This gene encodes a transcriptional repressor that interacts with several members of the BCL2 family of proteins. Overexpression of this protein induces apoptosis, which can be suppressed by co-expression of BCL2 proteins. The protein localizes to dot-like structures throughout the nucleus, and redistributes to a zone near the nuclear envelope in cells undergoing apoptosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: LLDRGNTRDKEASKEKGSEKGRAEGEWEDQEALDYFSDKESGKQKFNDSEGDDTEETEDYRQFRKSVLADQGKSFATASHRNTEEEGLKYKSKVSLKGNRESDGFREEKNYKLKETGYVVERPSTTKDKHKEED

Specifications

Antigen BCLAF1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant BCLAF1.
Dilution Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene BCLAF1
Gene Accession No. Q9NYF8
Gene Alias BTF/KIAA0164/bK211L9.1
Gene Symbols BCLAF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human BCLAF1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9774
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.