Learn More
BCLAF1 Rabbit anti-Chimpanzee, Dog, Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant BCLAF1.
Supplier: Abnova Corporation PAB0303
Description
This gene encodes a transcriptional repressor that interacts with several members of the BCL2 family of proteins. Overexpression of this protein induces apoptosis, which can be suppressed by co-expression of BCL2 proteins. The protein localizes to dot-like structures throughout the nucleus, and redistributes to a zone near the nuclear envelope in cells undergoing apoptosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
BCLAF1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant BCLAF1. | |
In PBS (0.2% gelatin, 0.05% sodium azide) | |
NP_055554 | |
BCLAF1 | |
Recombinant protein corresponding to amino acids 700-750 of human BCLAF1. | |
RUO | |
9774 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
BCLAF1 | |
BTF/KIAA0164/bK211L9.1 | |
Rabbit | |
100 μg | |
Primary | |
Canine, Chimpanzee, Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only