Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDKRB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89130129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-129 Supplier Abnova Corporation Supplier No. PAB28617

Rabbit polyclonal antibody raised against recombinant BDKRB1.

Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses. [provided by RefSeq

Sequence: MASSWPPLELQSSNQSQLSPQNATACDNAPEAWDLLHRVLP

Specifications

Antigen BDKRB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description bradykinin receptor B1
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene BDKRB1
Gene Accession No. G3V4Y2
Gene Alias B1BKR, B1R, BKB1R, BKR1, BRADYB1
Gene Symbols BDKRB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human BDKRB1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 623
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.