Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDNF Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114392 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-392 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-392 Supplier Abnova Corporation Supplier No. PAB0745
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BDNF.

The protein encoded by this gene is a member of the nerve growth factor family. It is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of this gene is reduced in both Alzheimer's and Huntington disease patients. This gene may play a role in the regulation of stress response and in the biology of mood disorders. Multiple transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]

Sequence: ELLDEDQKVRPNEE

Specifications

Antigen BDNF
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BDNF.
Dilution Immunohistochemistry (1-5 ug/mL) Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene BDNF
Gene Accession No. P23560
Gene Alias MGC34632
Gene Symbols BDNF
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 69-82 of human BDNF.
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 627
Test Specificity Used in western blot, this antiserum detects a 35 KDa band corresponding to the molecular weight of proBDNF. No cross reactivity with other proneurotrophins was detected.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.