Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BECN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114376 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-376 Supplier Abnova Corporation Supplier No. PAB0728
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BECN1.

Specifications

Antigen BECN1
Applications Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BECN1.
Dilution Immunohistochemistry (1:100-1:2000) Immunofluorescence (1:100-1:2000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene BECN1
Gene Accession No. Q14457
Gene Alias ATG6/VPS30/beclin1
Gene Symbols BECN1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human BECN1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8678
Test Specificity Specific for Beclin-1.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.