Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BECN1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115847 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-847 Supplier Abnova Corporation Supplier No. PAB12472
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BECN1.

Specifications

Antigen BECN1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BECN1.
Dilution Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene BECN1
Gene Accession No. Q14457
Gene Alias ATG6/VPS30/beclin1
Gene Symbols BECN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human BECN1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8678
Test Specificity This antibody is specific to human Beclin 1.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.