Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BECN1 Rabbit anti-Bovine, Chicken, Human, Mouse, Pig, Primates, Rat, Xenopus, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BECN1.
Supplier: Abnova Corporation PAB12473
Specifications
BECN1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BECN1. | |
In PBS (50% glycerol, 0.1% BSA, 0.05% sodium azide) | |
Q14457 | |
BECN1 | |
A synthetic peptide corresponding to amino acids 150-300 of human BECN1. | |
RUO | |
8678 | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunocytochemistry (1:50) Immunofluorescence (1:50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (1:10000) The optimal working dilution should be determined by the end user. | |
BECN1 | |
ATG6/VPS30/beclin1 | |
Rabbit | |
50 μL | |
Primary | |
Bovine, Chicken, Human, Mouse, Pig, Primate, Rat, Xenopus species | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction