Learn More
BEST2 Rabbit anti-Dog, Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BEST2.
Supplier: Abnova Corporation PAB27734
Description
This gene is a member of the bestrophin gene family of anion channels. Bestrophin genes share a similar gene structure with highly conserved exon-intron boundaries, but with distinct 3' ends. Bestrophins are transmembrane proteins that contain a homologous region rich in aromatic residues, including an invariant arg-phe-pro motif. Mutation in one of the family members (bestrophin 1) is associated with vitelliform macular dystrophy. The bestrophin 2 gene is mainly expressed in the retinal pigment epithelium and colon. [provided by RefSeq]
Specifications
BEST2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BEST2. | |
In PBS (0.09% sodium azide) | |
Q8NFU1 | |
BEST2 | |
A synthetic peptide corresponding to 15 amino acid at internal region of human BEST2. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins, except BEST1 (47%). | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5-5 ug/mL) The optimal working dilution should be determined by the end user. | |
BEST2 | |
FLJ20132/VMD2L1 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
54831 | |
Canine, Human, Monkey | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only